Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Polymerase epsilon p59 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2847970.1ML
Description
DNA Polymerase epsilon p59 Polyclonal specifically detects DNA Polymerase epsilon p59 in Human samples. It is validated for Western Blot.Specifications
DNA Polymerase epsilon p59 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
DNA polymerase epsilon subunit 2, DNA polymerase II subunit 2, DPE2DNA polymerase epsilon subunit B, EC 2.7.7.7, polymerase (DNA directed), epsilon 2 (p59 subunit) | |
The immunogen is a synthetic peptide directed towards the middle region of human DNA Polymerase epsilon p59 | |
0.1 mL | |
Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers | |
5427 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction