Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Polymerase epsilon subunit 3 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060438AF532
Description
DNA Polymerase epsilon subunit 3 Polyclonal antibody specifically detects DNA Polymerase epsilon subunit 3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DNA Polymerase epsilon subunit 3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Arsenic-transactivated protein, asTP, CHARAC17arsenic transactivated protein, CHRAC-17, CHRAC17Ybl1, Chromatin accessibility complex 17 kDa protein, DNA polymerase epsilon p17 subunit, DNA polymerase epsilon subunit 3, DNA polymerase epsilon subunit p17, DNA polymerase II subunit 3, EC 2.7.7.7, histone fold protein CHRAC17, HuCHRAC17, p17, polymerase (DNA directed), epsilon 3 (p17 subunit), YBL1 | |
The immunogen recognized by this antibody maps to a region between residue 97 and 147 of human DNA polymerase epsilon subunit 3 using the numbering given in entry NP_059139.2 (GeneID 54107). | |
0.1 mL | |
Chromatin Research, DNA Polymerases, DNA Repair | |
54107 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction