Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA polymerase eta Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982542
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2542 Supplier Bioss Supplier No. BS13016R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The XPA gene encodes a zinc metalloprotein that preferentially binds to DNA damaged by UV radiation and chemical carcinogens and is required for the incision step during nucleotide excision repair The XPB and XPD genes encode DNA helicases involved in several DNA metabolic pathways, including DNA repair and transcription, and the XPG gene product is an endonuclease that cuts on the 3′ side of a DNA lesion during nucleotide excision repair. Molecular defects in the XP variant (POL H) group maintain normal excision repair, yet they result in a substantial reduction in the ability to synthesize intact daughter DNA strands during DNA replication following DNA damage.

Specifications

Antigen DNA polymerase eta
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene POLH
Gene Accession No. Q9Y253
Gene Alias DNA polymerase eta; DNA-directed DNA polymerase eta; FLJ16395; FLJ21978; POLH; polymerase (DNA directed), eta; polymerase (DNA directed), eta (RAD 30 related); polymerase (DNA) eta; RAD30; RAD30 homolog A; RAD30A; Xeroderma pigmentosum variant type protein; Xeroderma pigmentosum variant type protein homolog; XP V; XPV; XP-V
Gene Symbols POLH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DNA polymerase eta.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5429
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.