Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Polymerase gamma Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306345APC
Description
DNA Polymerase gamma Polyclonal antibody specifically detects DNA Polymerase gamma in Human samples. It is validated for Western BlotSpecifications
DNA Polymerase gamma | |
Polyclonal | |
Western Blot | |
DNA polymerase subunit gamma-1, EC 2.7.7.7, FLJ27114, MDP1, MIRAS, Mitochondrial DNA polymerase catalytic subunit, MTDPS4A, PolG, catalytic subunit, POLG1MTDPS4B, PolG-alpha, POLGAPEO, polymerase (DNA directed), gamma, SANDO, SCAE | |
Produced in rabbits immunized with E. coli-derived Human DNA Polymerase gamma fragment. | |
0.1 mL | |
DNA Polymerases, DNA Repair | |
5428 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction