Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega DNA Polymerase I

Catalog No. PRM2051 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-M2051 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-M2051 Supplier Promega Supplier No. M2051
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNA Polymerase I catalyzes the template-directed polymerization of nucleotides into duplex DNA in a 5'→3' direction. Recombinant.

  • Catalyzes the Template-Directed Polymerization of Nucleotides into Duplex DNA in a 5′→3′ Direction
  • Can be used in a variety of molecular applications
  • Is inactivated by heating at 68°C for 10 minutes
  • Supplied at a concentration of 5-10u/μL

Specifications

Concentration 5 to 10 U/μL
Color Colorless
Components 10X Reaction Buffer: 500mM Tris HCl (pH 7.2 at 25°C), 100mM MgSO4, DTT 1mM
Purity >90%
pH 7.5
For Use With (Application) Labeling of DNA to high specific radioactivity by nick translation, second-strand cDNA synthesis
Storage Buffer 50mM Tris HCl (pH 7.5 at 25°C), 1mM DTT, 0.1mM EDTA, 50% Glycerol
Content And Storage -30°C to -10°C
Source Recombinant E. coli strain
Quantity 500 U
Product Type DNA Polymerase I
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.