Learn More
Thermo Scientific™ DNA Polymerase I (10 U/μL)
DNA Polymerase I is a template-dependent DNA polymerase which catalyzes 5'→3' synthesis of DNA.
$136.00 - $547.00
Specifications
Concentration | 10 U/μL |
---|---|
Description | DNA Polymerase I |
Polymerase | DNA Polymerase I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEREP0041
![]() |
Thermo Scientific™
EP0041 |
500 units |
Each for $136.00
|
|
|||||
FEREP0042
![]() |
Thermo Scientific™
EP0042 |
2,500 units |
Each for $547.00
|
|
|||||
Description
DNA Polymerase I, a template-dependent DNA polymerase, catalyzes 5'→3' synthesis of DNA. The enzyme also exhibits 3'→5' exonuclease (proofreading) activity, 5'→3' exonuclease activity, and ribonuclease H activity.
Highlights
- Incorporates modified nucleotides (e.g. biotin-, digoxigenin-, aminoallyl-, fluorescently-labeled nucleotides)
- Active in multiple buffers, including restriction enzyme, PCR, and RT buffers
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
10 U/μL | |
DNA Polymerase I | |
DNA Polymerase I |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.