Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Polymerase Kappa Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106559AF350
Description
DNA Polymerase Kappa Polyclonal antibody specifically detects DNA Polymerase Kappa in Human samples. It is validated for Western BlotSpecifications
DNA Polymerase Kappa | |
Polyclonal | |
Western Blot | |
DINB protein, DINPPOLQDINB1EC 2.7.7.7, DNA polymerase kappa, polymerase (DNA directed) kappa | |
The immunogen recognized by this antibody maps to a region between residue 825 and 870 of human DNA polymerase kappa using the numbering given in entry NP_057302.1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
51426 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction