Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase Kappa/POLK Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982545
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2545 Supplier Bioss Supplier No. BS13019R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

External and internal DNA-damaging agents continually threaten the integrity of genetic material in cells. Although a variety of repair mechanisms exist to remove the resulting lesions, some lesions escape repair and block the replication machinery. Members of the Y family of DNA polymerases, such as POLK, permit the continuity of the replication fork by allowing replication through such DNA lesions. Each Y family polymerase has a unique DNA-damage bypass and fidelity profile. POLK is specialized for the extension step of lesion bypass.

Specifications

Antigen DNA Polymerase Kappa/POLK
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene POLK
Gene Accession No. Q9UBT6
Gene Alias BOS_10072; DinB homolog 1; DINB protein; DINB1; DINP; DNA damage-inducible protein b; DNA polymerase kappa; DNA-directed DNA polymerase kappa; Polk; POLQ; polymerase (DNA directed) kappa; polymerase (DNA directed), kappa; polymerase (DNA) kappa
Gene Symbols POLK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DNA Polymerase Kappa/POLK.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51426
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.