Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA polymerase mu Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982547
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2547 Supplier Bioss Supplier No. BS13021R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gap-filling polymerase involved in repair of DNA double-strand breaks by non-homologous end joining (NHEJ). Participates in immunoglobulin (Ig) light chain gene rearrangement in V(D)J recombination.

Specifications

Antigen DNA polymerase mu
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene POLM
Gene Accession No. Q9NP87
Gene Alias DNA polymerase mu; DNA-directed DNA/RNA polymerase mu; Pol iota; Pol Mu; POLM; polmu; polymerase (DNA directed), mu; polymerase (DNA) mu; polymerase (DNA-directed), mu; Tdt-N; Terminal transferase
Gene Symbols POLM
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DNA polymerase mu.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27434
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.