Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA polymerase sigma Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen DNA polymerase sigma
Dilution Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056356T
SDP
View Documents
Novus Biologicals
NB10056356SS
0.025 mL
Each for $181.90
Only null left
Add to Cart
 
NB10056356
SDP
View Documents
Novus Biologicals
NB10056356
0.1 mL
Each for $402.00
Only null left
Add to Cart
 
Description

Description

DNA polymerase sigma Polyclonal specifically detects DNA polymerase sigma in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

DNA polymerase sigma
Western Blot
Unconjugated
Rabbit
Human
NP_008930.1
11044
This antibody was generated by immunizing rabbits with a synthetic peptide containing amino acids 447-462 (GTASLKAVHHMSSPAI) of human TRF4-1, GenBank no NP_008930.1.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500
Polyclonal
Purified
RUO
PBS with 0.5% BSA with 0.02% Sodium Azide
DNA polymerase kappa, DNA polymerase sigma, EC 2.7.7.7, LAK-1TUTase 5, PAP associated domain containing 7, POLKTRF41, POLSTUTASE5, polymerase (DNA directed) sigma, polymerase (DNA-directed) sigma, Terminal uridylyltransferase 5, Topoisomerase-related function protein 4-1, TRF4-1LAK1, TRF4PAP-associated domain-containing protein 7
TENT4A
IgG
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.