Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA polymerase subunit 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988042
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8042 Supplier Bioss Supplier No. BS7823R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a zinc finger protein that may act as a transcription factor. The encoded protein may be part of a BHC histone deacetylase complex. Translocation of this gene with the fibroblast growth factor receptor-1 gene (FGFR1) results in a fusion gene, which may be a cause of stem cell leukemia lymphoma syndrome (SCLL). Several transcript variants encoding the same protein have been found for this gene.

Specifications

Antigen DNA polymerase subunit 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene POLA2
Gene Accession No. Q14181
Gene Alias AI573378; BOS_24809; DNA polymerase alpha 2, 68 kDa; DNA polymerase alpha 2, accessory subunit; DNA polymerase alpha 70 kDa subunit; DNA polymerase alpha subunit B; DNA polymerase alpha subunit II; DNA polymerase subunit II; DNA-directed DNA polymerase alpha 2; Ese1L; Pola2; polymerase (DNA directed), alpha 2; polymerase (DNA directed), alpha 2 (70kD subunit); polymerase (DNA directed), alpha 2, accessory subunit; polymerase (DNA) alpha 2, accessory subunit; polymerase (DNA-directed), alpha (70kD)
Gene Symbols POLA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human POLA2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23649
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.