Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNA Primase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550022
Description
The antibody detects endogenous levels of total DNA Primase protein.
The replication of DNA in eukaryotic cells is carried out by a complex chromosomal replication apparatus, in which DNA polymerase alpha and primase are two key enzymatic components. Primase, which is a heterodimer of a small subunit and a large subunit, synthesizes small RNA primers for the Okazaki fragments made during discontinuous DNA replication. The protein encoded by this gene is the small, 49 kDa primase subunit.
Specifications
DNA Primase | |
Polyclonal | |
Unconjugated | |
PRIM1 | |
AI324982; DNA primase 1; DNA primase 49 kDa subunit; DNA primase small subunit; DNA primase subunit 48; DNA primase, p49 subunit; MGC12308; p49; PRI1; PRIM1; primase (DNA) subunit 1; primase p49 subunit; primase polypeptide 1, 49kDa; primase, DNA, polypeptide 1; primase, DNA, polypeptide 1 (49kDa) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19075, 246327, 5557 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P20664, P49642 | |
PRIM1 | |
Synthesized peptide derived from C-terminal of human PRIM1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction