Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA Primase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559027
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559027 Supplier Invitrogen™ Supplier No. PA559027
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELVFDIDMTD YDDVRRCCSS ADICPKCWTL MTMAIRIIDR ALKEDFGFKH RLWVYSGRRG VHCWVCDESV RKLSSAVRSG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 74%.

The replication of DNA in eukaryotic cells is carried out by a complex chromosomal replication apparatus, in which DNA polymerase alpha and primase are two key enzymatic components. Primase, which is a heterodimer of a small subunit and a large subunit, synthesizes small RNA primers for the Okazaki fragments made during discontinuous DNA replication. The protein encoded by this gene is the small, 49 kDa primase subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Primase
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRIM1
Gene Accession No. P49642
Gene Alias AI324982; DNA primase 1; DNA primase 49 kDa subunit; DNA primase small subunit; DNA primase subunit 48; DNA primase, p49 subunit; MGC12308; p49; PRI1; PRIM1; primase (DNA) subunit 1; primase p49 subunit; primase polypeptide 1, 49kDa; primase, DNA, polypeptide 1; primase, DNA, polypeptide 1 (49kDa)
Gene Symbols PRIM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNA Primase. Recombinant protein control fragment (Product #RP-97891).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5557
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.