Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA protection during starvation protein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117548
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117548 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117548 Supplier Invitrogen™ Supplier No. PA5117548
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Protects DNA from oxidative damage by sequestering intracellular Fe(2+) ion and storing it in the form of Fe(3+) oxyhydroxide mineral. One hydrogen peroxide oxidizes two Fe(2+) ions, which prevents hydroxyl radical production by the Fenton reaction. It protects DNA from hydroxyl radical-mediated cleavage. Binds DNA with no apparent sequence specificity without self-aggregation nor promotion of DNA condensation. Is unable to protect DNA from DNase-mediated cleavage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA protection during starvation protein
Applications Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene dps
Gene Accession No. P0C558
Gene Symbols dps
Host Species Rabbit
Immunogen Recombinant Mycobacterium smegmatis DNA protection during starvation protein (1-183aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 45747735
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.