Learn More
DNA/RNA Damage Antibody (15A3), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NB11096878FR
Catalog No. NBT96878FR
Only null left
Add to Cart
Description
Description
DNA/RNA Damage Monoclonal specifically detects DNA/RNA Damage in Human, Mouse, Non-species specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
DNA/RNA Damage
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
8-hydroxy-guanosine-BSA and-casein conjugates
0.1 mL
Recognizes markers of oxidative damage to DNA (8-hydroxy-2'-deoxyguanosine, 8-hydroxyguanine and 8-hydroxyguanosine).
Store at 4C in the dark.
IgG2b
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
15A3
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Mouse
Protein G purified
Primary
Human, Mouse, Non-species specific
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.