Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA/RNA Damage Antibody (15A3), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11096878MFV450
Description
DNA/RNA Damage Monoclonal antibody specifically detects DNA/RNA Damage in Human, Mouse, Non-species specific samples. It is validated for Flow CytometrySpecifications
DNA/RNA Damage | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Human, Mouse, Non-species specific | |
Purified |
Flow Cytometry | |
15A3 | |
Flow Cytometry | |
DNA/RNA Oxidative Damage Markers Monoclonal | |
8-hydroxy-guanosine-BSA and-casein conjugates | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction