Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA-RNA Hybrid Monoclonal Antibody (S9.6), Active Motif™
SDP

Catalog No. 501992613
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2613 100 μg
50-199-2614 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2613 Supplier Active Motif Supplier No. 65683
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

DNA-RNA hybrids occur naturally during cellular functions such as transcription and replication. However, they may be a significant source of genome instability leading to cancer and other genetic diseases. Detection of DNA-RNA hybrids may lead to the discovery of new disease targets.

Specifications

Antigen DNA-RNA Hybrid
Applications Dot Blot
Classification Monoclonal
Clone S9.6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Host Species Mouse
Immunogen Propietary
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Multi-species, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.