Learn More
G-Biosciences DNA Size Selection Magnetic Beads
SCI_ED
DNA Size Selection Magnetic Beads are suitable for DNA fragment size selection from 150 bp to 1000 bp and even larger.
$231.00 - $5381.00
Specifications
Product Type Magnetic Beads
Primer Random
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S04209
SCI_ED
View Documents
G-Biosciences
7862158
5 mL
Each for $231.00
Only null left
Add to Cart
S04224
SCI_ED
View Documents
G-Biosciences
7862159
60 mL
Each for $1,384.00
Only null left
Add to Cart
S04225
SCI_ED
View Documents
G-Biosciences
7862160
450 mL
Each for $5,381.00
Only null left
Add to Cart
Description
Description
DNA size selection Magnetic Beads remove excess nucleotides, salts, enzymes, and other impurities with a simple washing process to obtain purified fragments, which can be directly used in downstream applications such as sequencing, hybridization, and enzyme digestion. DNA size selection depends on optimizing magnetic beads, target DNA ratio, and salt concentrations.
Specifications
Specifications
Magnetic Beads
Random
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.