Learn More
G-Biosciences DNA Size Selection and PCR Clean Up Magnetic Beads
SCI_ED
Catalog No. S04208
Change view
Click to view available options
Quantity:
5 mL
60 mL
450 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04208 5 mL
S04226 60 mL
S04218 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S04208 Supplier G-Biosciences Supplier No. 7862164
Add to Cart
Add to Cart
G-Biosciences DNA Size Selection and PCR Cleanup Magnetic Beads are suitable for PCR clean-up and DNA fragment size selection from 150 bp to 1000 bp and even larger.
DNA size selection and PCR clean up Magnetic Beads remove excess nucleotides, salts, enzymes, and other impurities with a simple washing process to obtain purified fragments, which can be directly used in downstream applications such as sequencing, hybridization, and enzyme digestion. DNA size selection depends on optimizing magnetic beads, target DNA ratio, and salt concentrations.
Specifications
Product Type Magnetic Beads
Primer Random
Quantity 5 mL
Product Line Ambion, Applied Biosystems, DNAZap, RNaseAlert, RNase Away