Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568743 Supplier Invitrogen™ Supplier No. PA568743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 93%; Guinea Pig: 85%; Horse: 86%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 85%; Rat: 86%.

DNA2 is a conserved helicase/nuclease involved in themaintenance of mitochondrial and nuclear DNA stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DNA2
Gene Accession No. P51530
Gene Alias DNA replication ATP-dependent helicase DNA2; DNA replication ATP-dependent helicase/nuclease DNA2; DNA replication ATP-dependent helicase-like homolog; DNA replication helicase 2 homolog; DNA replication helicase 2 homolog (yeast); DNA replication helicase 2-like; DNA replication helicase/nuclease 2; DNA replication nuclease DNA2; DNA2; DNA2L; DNA2-like helicase; E130315B21Rik; hDNA2; Kiaa0083; LOW QUALITY PROTEIN: DNA replication ATP-dependent helicase/nuclease DNA2
Gene Symbols DNA2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human DNA2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1763
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.