Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118398 Supplier Invitrogen™ Supplier No. PA5118398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNA2 Polyclonal Antibody for Flow, IHC (P)

DNA2 is a conserved helicase/nuclease involved in themaintenance of mitochondrial and nuclear DNA stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA2
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene DNA2
Gene Accession No. D3ZG52, P51530, Q6ZQJ5
Gene Alias DNA replication ATP-dependent helicase DNA2; DNA replication ATP-dependent helicase/nuclease DNA2; DNA replication ATP-dependent helicase-like homolog; DNA replication helicase 2 homolog; DNA replication helicase 2 homolog (yeast); DNA replication helicase 2-like; DNA replication helicase/nuclease 2; DNA replication nuclease DNA2; DNA2; DNA2L; DNA2-like helicase; E130315B21Rik; hDNA2; Kiaa0083; LOW QUALITY PROTEIN: DNA replication ATP-dependent helicase/nuclease DNA2
Gene Symbols DNA2
Host Species Rabbit
Immunogen Synthetic peptide within human DNA2 aa 960-1000.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1763, 309762, 327762
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.