Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501733505 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3505 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3505 Supplier Proteintech Group Inc Supplier No. 215991AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specific to DNA2.

DNA2 is a conserved helicase/nuclease involved in themaintece of mitochondrial and nuclear DNA stability.

Specifications

Antigen DNA2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DNA2
Gene Accession No. P51530
Gene Alias DNA2, DNA2L, KIAA0083
Gene Symbols DNA2
Host Species Rabbit
Immunogen DNA2 Fusion Protein Ag16206
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1763
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.