Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNAAF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551766
Description
Immunogen sequence: LQGKEERVNE ESHLTEKEYI EHCNTPTTDS DSSIAVKALQ IDSFGLVTCF QQESLDVSQM ILGKSQQPES KMQSEFIKEK SATCSNEEKG NLNESVITEE KETDGDHLSS LLNKTTVHNI PGFDSIKETN MQDGSVQVIK Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 57%.
The function of this protein remains unknown.
Specifications
| DNAAF2 | |
| Polyclonal | |
| Unconjugated | |
| DNAAF2 | |
| 1110034A24Rik; 2810020C19Rik; AV032410; C10H14orf104; C14orf104; CILD10; DNAAF2; dynein (axonemal) assembly factor 2; dynein assembly factor 2, axonemal; dynein axonemal assembly factor 2; dynein, axonemal assembly factor 2; dynein, axonemal, assembly factor 2; kintoun; kintoun homolog; Ktu; mknt; PF13; protein kintoun; RGD1310311; TU54 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55172 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NVR5 | |
| DNAAF2 | |
| Recombinant protein corresponding to Human DNAAF2. Recombinant protein control fragment (Product #RP-89061). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction