Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAAF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544697
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544697 Supplier Invitrogen™ Supplier No. PA544697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MFSQYAEELT DPENRRRYEA EITALERERG VEVRFVHPEP GHVLRTSLDG Sequence homology: Cow: 100%; Dog: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAAF2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DNAAF2
Gene Accession No. Q9NVR5
Gene Alias 1110034A24Rik; 2810020C19Rik; AV032410; C10H14orf104; C14orf104; CILD10; DNAAF2; dynein (axonemal) assembly factor 2; dynein assembly factor 2, axonemal; dynein axonemal assembly factor 2; dynein, axonemal assembly factor 2; dynein, axonemal, assembly factor 2; kintoun; kintoun homolog; Ktu; mknt; PF13; protein kintoun; RGD1310311; TU54
Gene Symbols DNAAF2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C14orf104 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55172
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.