Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAAF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566004 Supplier Invitrogen™ Supplier No. PA566004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FVLENNLEAVA RHMLIFSLAL EEPEKMGLQE RSETFLEVWG NALLRPPVAA FVRAQADLLA HLVPEPDRLE EQLPWLSLRA LKFRERDALE AVFRFWA Highest antigen sequence identity to the following orthologs - mouse 90%, rat 89%.

The protein encoded by this gene is required for the assembly of axonemal inner and outer dynein arms and plays a role in assembling dynein complexes for transport into cilia. Defects in this gene are a cause of primary ciliary dyskinesia type 2 (CILD2). Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAAF3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAAF3
Gene Accession No. Q8N9W5
Gene Alias 6030429G01Rik; b2b1739Clo; C19orf51; CILD2; DAB1; DNAAF3; dynein; dynein (axonemal) assembly factor 3; dynein assembly factor 3, axonemal; dynein axonemal assembly factor 3; dynein, axonemal assembly factor 3; dynein, axonemal, assembly factor 3; PCD; PF22; UPF0470 protein C19orf51; UPF0470 protein C19orf51 homolog
Gene Symbols DNAAF3
Host Species Rabbit
Immunogen Recombinant Human DNAAF3. Recombinant protein control fragment (Product #RP-106691).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 352909
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.