Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNAAF3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571524
Description
DNAAF3 Polyclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is required for the assembly of axonemal inner and outer dynein arms and plays a role in assembling dynein complexes for transport into cilia. Defects in this gene are a cause of primary ciliary dyskinesia type 2 (CILD2). Several transcript variants encoding different isoforms have been found for this gene.
Specifications
DNAAF3 | |
Polyclonal | |
Unconjugated | |
DNAAF3 | |
6030429G01Rik; b2b1739Clo; C19orf51; CILD2; DAB1; DNAAF3; dynein; dynein (axonemal) assembly factor 3; dynein assembly factor 3, axonemal; dynein axonemal assembly factor 3; dynein, axonemal assembly factor 3; dynein, axonemal, assembly factor 3; PCD; PF22; UPF0470 protein C19orf51; UPF0470 protein C19orf51 homolog | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
352909, 436022 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q3UYV8, Q8N9W5 | |
DNAAF3 | |
KLH conjugated synthetic peptide between 181-210 amino acids from the Central region of human C19orf51. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction