Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557826
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557826 Supplier Invitrogen™ Supplier No. PA557826
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RIVKVMDDYQ VMDEFLYNLS SDDFNDKWIA SNWPSKILGQ IELVQQQHVE DEEKFRKIQI MDQNNFQEKL EGLQLVVAGF S Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.

This gene encodes an inner dynein arm heavy chain that provides structural support between the radial spokes and the outer doublet of the sperm tail. Naturally occurring mutations in this gene are associated with primary ciliary dyskinesia and multiple morphological anomalies of the flagella that result in asthenozoospermia and male infertility. Mice with a homozygous knockout of the orthologous gene are viable but have reduced sperm motility and are infertile.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAH1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAH1
Gene Accession No. Q9P2D7
Gene Alias axonemal beta dynein heavy chain 1; Ciliary dynein heavy chain 1; DHC7; Dlp1; DNAH1; DNAH1 variant protein; Dnahc1; dynein axonemal heavy chain 1; dynein heavy chain 1, axonemal; dynein, axonemal, heavy chain 1; dynein, axonemal, heavy polypeptide 1; hDHC7; Heat shock regulated protein 1; HL11; HL-11; HSRF-1; KIAA1410; RGD1311110; testicular tissue protein Li 60; XLHSRF-1
Gene Symbols DNAH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAH1. Recombinant protein control fragment (Product #RP-96323).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25981
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.