Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAH10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558645 Supplier Invitrogen™ Supplier No. PA558645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRLTLIEAIN LFKYPAAKSE EELPGVKEFF EHIERERASD VDHMVRWYLA IGPLLTKVEG LVVHTNTGKA PKLASYYKYW EKKIYEVLTK LILKNLQ Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 20%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAH10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAH10
Gene Accession No. Q8IVF4
Gene Alias Axonemal beta dynein heavy chain 10; ciliary dynein heavy chain 10; DNAH10; Dnahc10; dynein axonemal heavy chain 10; dynein heavy chain 10, axonemal; dynein, axon, heavy chain 10; dynein, axonemal, heavy chain 10; dynein, axonemal, heavy polypeptide 10; KIAA2017
Gene Symbols DNAH10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAH10. Recombinant protein control fragment (Product #RP-97884).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196385
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.