Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAH12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23891625UL
Description
DNAH12 Polyclonal specifically detects DNAH12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DNAH12 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Q6ZR08 | |
DNAH12 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LLLTMLADFYNLYIVENPHYKFSPSGNYFAPPKGTYEDYIEFIKKLPFTQHP | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
axonemal beta dynein heavy chain 12, axonemal dynein heavy chain isotype3, axonemal, heavy polypeptide 12, ciliary dynein heavy chain 12, DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, dynein heavy chain 12, axonemal, dynein heavy chain domain-containing protein 2, dynein, axonemal, heavy chain 12, dynein, heavy chain-5, FLJ40427, FLJ44290, HDHC3, HL19, HL-19 | |
Rabbit | |
Affinity Purified | |
RUO | |
201625 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction