Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAH12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570821 Supplier Invitrogen™ Supplier No. PA570821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 100%; Horse: 93%; Human: 100%; Rabbit: 93%; Rat: 86%.

DNAH12 gene ontology annotations related to this gene include microtubule-based movement.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAH12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DNAH12
Gene Accession No. Q6ZR08
Gene Alias 4921531P07Rik; axonemal beta dynein heavy chain 12; Axonemal dynein heavy chain 12-like protein; Axonemal dynein heavy chain 7-like protein; axonemal dynein heavy chain isotype3; Bm259; ciliary dynein heavy chain 12; DHC3; DLP12; Dnah12; DNAH12L; DNAH7L; DNAHC12; Dnahc3; Dnahc7c; Dnahc7l; DNHD2; dynein axonemal heavy chain 12; dynein heavy chain 12, axonemal; dynein heavy chain 12, axonemal-like; Dynein heavy chain 7-like, axonemal; dynein heavy chain domain 2; dynein heavy chain domain-containing protein 2; dynein, axonemal, heavy chain 12; dynein, axonemal, heavy polypeptide 12; dynein, heavy chain-5; Gm284; Gm74; Gm907; Hdhc3; HL19; HL-19
Gene Symbols DNAH12
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human DNAH12.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201625
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.