Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAH17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584995 Supplier Invitrogen™ Supplier No. PA584995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PWKDYVIYID DMVLDEFDQF IRKSLSFLMD NMVIDESIAP LFEIRMELDE DGLTFNPTLE VGSDRGFLAL IEGLVNDIYN VARLIPRLAK DRMNYKMD Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 78%.

Dyneins are microtubule-associated motor protein complexes composed of several heavy, light, and intermediate chains. DNAH17 is a heavy chain associated with axonemal dynein (Milisav and Affara, 1998 [PubMed 9545504]).[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAH17
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAH17
Gene Accession No. Q9UFH2
Gene Alias Axonemal beta dynein heavy chain 17; axonemal dynein heavy chain; axonemal dynein heavy chain-like protein 1; Ciliary dynein heavy chain 17; ciliary dynein heavy chain-like protein 1; DNAH17; DNAHL1; DNEL2; dynein axonemal heavy chain 17; dynein heavy chain 17, axonemal; Dynein light chain 2, axonemal; dynein, axonemal, heavy chain 17; dynein, axonemal, heavy chain like 1; dynein, axonemal, heavy like 1; dynein, axonemal, heavy polypeptide 17; FLJ40457
Gene Symbols DNAH17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAH17. Recombinant protein control fragment (Product #RP-108279).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8632
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.