Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564309
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564309 Supplier Invitrogen™ Supplier No. PA564309
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGEWENACNE MQRMLIVRSL RQDRVAFCVT SFIITNLGSR FIEPPVLNMK SVLEDSTPRS PLVFILSPGV DPTSALL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

Force generating protein of respiratory cilia. Produces force towards the minus ends of microtubules. Dynein has ATPase activity; the force-producing power stroke is thought to occur on release of ADP. Involved in sperm motility; implicated in sperm flagellar assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAH2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAH2
Gene Accession No. Q9P225
Gene Alias axonemal beta dynein heavy chain 2; Ciliary dynein heavy chain 2; DNAH2; DNAHC2; DNHD3; dynein axonemal heavy chain 2; dynein heavy chain 2, axonemal; Dynein heavy chain domain-containing protein 3; dynein, axonemal, heavy chain 2; dynein, axonemal, heavy polypeptide 2; KIAA1503
Gene Symbols DNAH2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAH2. Recombinant protein control fragment (Product #RP-103886).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146754
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.