Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAH5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557958 Supplier Invitrogen™ Supplier No. PA557958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVEDAILEGN QIERIDQLFA VGGLRHLMFY YQDVEEAETG QLGSLGGVNL VSGKIKKPKV FVTEGNDVAL TGVCVFFIRT DPSKAITPD Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

This gene encodes a dynein protein, which is part of a microtubule-associated motor protein complex consisting of heavy, light, and intermediate chains. This protein is an axonemal heavy chain dynein. It functions as a force-generating protein with ATPase activity, whereby the release of ADP is thought to produce the force-producing power stroke. Mutations in this gene cause primary ciliary dyskinesia type 3, as well as Kartagener syndrome, which are both diseases due to ciliary defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAH5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAH5
Gene Accession No. Q8TE73
Gene Alias AU022615; Axonemal beta dynein heavy chain 5; axonemal dynein heavy chain 5; b2b1134Clo; b2b1154Clo; b2b1537Clo; b2b1565Clo; b2b601Clo; CILD3; Ciliary dynein heavy chain 5; DLP5; Dnah5; Dnahc5; Dnahc8; dynein axonemal heavy chain 5; dynein heavy chain 5, axonemal; dynein, axon, heavy chain 5; dynein, axonemal, heavy chain 5; dynein, axonemal, heavy polypeptide 5; dynein-like protein 5; HL1; KIAA1603; KTGNR; Mdnah5; mKIAA1603; PCD
Gene Symbols DNAH5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAH5. Recombinant protein control fragment (Product #RP-96796).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1767
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.