Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAH6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570137 Supplier Invitrogen™ Supplier No. PA570137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNAH6 Polyclonal Antibody for Western Blot

DNAH6 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAH6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DNAH6
Gene Accession No. Q9C0G6
Gene Alias 9830168K20; A730004I20Rik; axo a; Axonemal beta dynein heavy chain 6; axonemal dynein heavy chain; ciliary dynein heavy chain 6; DNAH6; Dnahc6; DNHL1; dynein axonemal heavy chain 6; dynein heavy chain 6, axonemal; dynein heavy chain-like 1; dynein, axon, heavy chain 6; dynein, axonemal, heavy chain 6; dynein, axonemal, heavy polypeptide 6; HL2; HL-2; KIAA1697; mdhc6; mKIAA1697
Gene Symbols DNAH6
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human DNAH6.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1768
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.