Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAH9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545744 Supplier Invitrogen™ Supplier No. PA545744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DSQARDGAGA TREEKVKALL EEILERVTDE FNIPELMAKV EERTPYIVVA Sequence homology: Cow: 83%; Dog: 100%; Guinea Pig: 83%; Horse: 92%; Human: 100%; Mouse: 92%; Rat: 92%.

This gene encodes the heavy chain subunit of axonemal dynein, a large multi-subunit molecular motor. Axonemal dynein attaches to microtubules and hydrolyzes ATP to mediate the movement of cilia and flagella. The gene expresses at least two transcript variants; additional variants have been described, but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAH9
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DNAH9
Gene Accession No. Q9NYC9
Gene Alias 9030022M04; A230091C01; axonemal beta dynein heavy chain 9; Bv2; C230051G16; ciliary dynein heavy chain 9; D11Ertd686e; DNAH17L; Dnah9; DNAH9 variant protein; Dnahc9; DNAL1; DNEL1; DYH9; dynein axonemal heavy chain 9; dynein heavy chain; dynein heavy chain 9, axonemal; dynein heavy chain Bv2; dynein, axonemal, heavy chain 9; dynein, axonemal, heavy polypeptide 9; HL20; HL-20; KIAA0357; mKIAA0357
Gene Symbols DNAH9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human DNAH9 (aa 4206-4255).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1770, 237806
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.