Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAI1/2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981039
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1039 Supplier Bioss Supplier No. BS11024R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Eukaryotic cells rely on actin and microtubule-based protein ''motors'' to generate intracellular movements. These protein ''motors'' contain specialized domains that hydrolyze ATP to produce force and movement along a cytoskeletal polymer (actin in the case of myosin family and microtubules in the case of the kinesin family and dyneins). Dynein has been implicated in cytoplasmic motile functions, including chromosomal movement, retrograde organelle and axonal transport, the endocytic pathway, and the organization of the Golgi apparatus. In all cell types, dynein has the same basic structures and is composed of two or three distinct heavy chains (˜450 kDa), three intermediate chains (70-125 kDa), and at least four light chains (15-25 kDa).

Specifications

Antigen DNAI1/2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DNAI1
Gene Accession No. Q9UI46
Gene Alias Axonemal dynein intermediate chain 1; CILD1; DIC1; DNAI1; Dnaic1; dynein axonemal intermediate chain 1; Dynein intermediate chain 1, axonemal; dynein, axonemal, intermediate chain 1; dynein, axonemal, intermediate polypeptide 1; I79_025143; ICS; ICS1; immotile cilia syndrome 1; MGC26204; PCD; RP11-296L22.2; testis tissue sperm-binding protein Li 87P
Gene Symbols DNAI1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DNAI1/2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27019
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.