Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAI1 Monoclonal Antibody (70.1)
GREENER_CHOICE

Catalog No. MA122146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA122146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA122146 Supplier Invitrogen™ Supplier No. MA122146
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: chicken brain. This antibody is know to detect cytoplasmic Dynein intermediate chains, and reactivity with the axonemal form is not known. By Western blot, MA1-22146 detects a ~ 70 kDa triplet representing the Dynein intermediate chains. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Eukaryotic cells rely on actin and microtubule-based protein 'motors' to generate intracellular movements. These protein 'motors' contain specialized domains that hydrolyze ATP to produce force and movement along a cytoskeletal polymer (actin in the case of myosin family and microtubules in the case of the kinesin family and dyneins). Dynein has been implicated in cytoplasmic motile functions, including chromosomal movement, retrograde organelle and axonal transport, the endocytic pathway, and the organization of the Golgi apparatus. In all cell types, dynein has the same basic structures and is composed of two or three distinct heavy chains (~450 kDa), three intermediate chains (70-125 kDa), and at least four light chains (15-25 kDa).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAI1
Applications ELISA, Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 70.1
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation Ascites with 15mM sodium azide
Gene DNAI1
Gene Accession No. Q5XIL8, Q9UI46
Gene Alias 1110066F04Rik; axonemal dynein intermediate chain 1; b2b1526Clo; BB124644; CILD1; DIC1; Dnai1; Dnaic1; dynein axonemal intermediate chain 1; Dynein intermediate chain 1, axonemal; dynein, axonemal, intermediate chain 1; dynein, axonemal, intermediate polypeptide 1; I79_025143; ICS; ICS1; immotile cilia syndrome 1; MGC26204; PCD; RP11-296L22.2; testis tissue sperm-binding protein Li 87P
Gene Symbols DNAI1
Host Species Mouse
Immunogen Cytoplasmic dynein from chicken embryo brain.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100754021, 27019, 431654, 500442
Target Species Human, Mouse, Rat, Hamster, Chicken, Amphibian, Marsupial, Deer
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.