Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 127561AP150UL

Description
Eukaryotic cells rely on actin and microtubule-based protein ""motors"" to generate intracellular movements. These protein ""motors"" contain specialized domains that hydrolyze ATP to produce force and movement along a cytoskeletal polymer (actin in the case of myosin family and microtubules in the case of the kinesin family and dyneins). Dynein has been implicated in cytoplasmic motile functions, including chromosomal movement, retrograde organelle and axonal transport, the endocytic pathway, and the organization of the Golgi apparatus. In all cell types, dynein has the same basic structures and is composed of two or three distinct heavy chains (∽450 kDa), three intermediate chains (70-125 kDa), and at least four light chains (15-25 kDa).Specifications
DNAI1 | |
Polyclonal | |
Unconjugated | |
DNAI1 | |
CILD1, DNAI1, ICS, ICS1, PCD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27019, 500442, 68922 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5XIL8, Q8C0M8, Q9UI46 | |
Dnai1 | |
DNAI1 Fusion Protein Ag3473 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction