Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAI1 Recombinant Rabbit Monoclonal Antibody (4Y6Q1)
GREENER_CHOICE

Catalog No. PIMA542832
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542832 Supplier Invitrogen™ Supplier No. MA542832
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: Mouse lung, Mouse testis, Rat testis. Immunogen sequence: MIPASAKAPH KQPHKQSISI GRGTRKRDED SGTEVGEGTD EWAQSKATVR PPDQLELTDA ELKEEFTRIL TANNPHAPQN IVRYSFKEGT YKPIGFVNQL AVHYTQVGNL IPKDSDEGRR QHYRDELVAG SQESVKVISE TGNLEEDEEP KELETEPGSQ TDVPAAGAAE KVTEEELMTP KQPKERKLTN QFNFSERASQ.

Eukaryotic cells rely on actin and microtubule-based protein 'motors' to generate intracellular movements. These protein 'motors' contain specialized domains that hydrolyze ATP to produce force and movement along a cytoskeletal polymer (actin in the case of myosin family and microtubules in the case of the kinesin family and dyneins). Dynein has been implicated in cytoplasmic motile functions, including chromosomal movement, retrograde organelle and axonal transport, the endocytic pathway, and the organization of the Golgi apparatus. In all cell types, dynein has the same basic structures and is composed of two or three distinct heavy chains (~450 kDa), three intermediate chains (70-125 kDa), and at least four light chains (15-25 kDa).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAI1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4Y6Q1
Concentration 1.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DNAI1
Gene Accession No. Q5XIL8
Gene Alias 1110066F04Rik; axonemal dynein intermediate chain 1; b2b1526Clo; BB124644; CILD1; DIC1; Dnai1; Dnaic1; dynein axonemal intermediate chain 1; Dynein intermediate chain 1, axonemal; dynein, axonemal, intermediate chain 1; dynein, axonemal, intermediate polypeptide 1; I79_025143; ICS; ICS1; immotile cilia syndrome 1; MGC26204; PCD; RP11-296L22.2; testis tissue sperm-binding protein Li 87P
Gene Symbols DNAI1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human Dynein intermediate chain 1 (Q9UI46).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500442
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.