Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101565
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101565 Supplier Invitrogen™ Supplier No. PA5101565
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DNAI2.

The protein encoded by this gene belongs to the dynein intermediate chain family, and is part of the dynein complex of respiratory cilia and sperm flagella. Mutations in this gene are associated with primary ciliary dyskinesia type 9. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAI2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DNAI2
Gene Accession No. A2AC93, Q66HC9, Q9GZS0
Gene Alias axonemal dynein intermediate chain 2; C030015H18Rik; CILD9; DIC2; Dnai2; Dnaic2; dynein axonemal intermediate chain 2; dynein intermediate chain 2, axonemal; dynein, axonemal, intermediate chain 2; dynein, axonemal, intermediate polypeptide 2; Gm728
Gene Symbols DNAI2, Dnaic2
Host Species Rabbit
Immunogen A synthesized peptide derived from human DNAI2(Accession Q9GZS0), corresponding to amino acid residues R72-K122.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360654, 432611, 64446
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.