Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562215
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562215 Supplier Invitrogen™ Supplier No. PA562215
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IVYVYVKKRS EFGKQCNFSD RQAELNIDIM PNPELAEQFV ERNPVDTGIQ CSISMSEHEA NSERFEMETR GVNHVEGGWP KDVNPLELEQ TIRFRKKVEK DENYVNAIMQ LGSIMEHCIK QNNAI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

The protein encoded by this gene belongs to the dynein intermediate chain family, and is part of the dynein complex of respiratory cilia and sperm flagella. Mutations in this gene are associated with primary ciliary dyskinesia type 9. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAI2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAI2
Gene Accession No. Q9GZS0
Gene Alias axonemal dynein intermediate chain 2; C030015H18Rik; CILD9; DIC2; Dnai2; Dnaic2; dynein axonemal intermediate chain 2; dynein intermediate chain 2, axonemal; dynein, axonemal, intermediate chain 2; dynein, axonemal, intermediate polypeptide 2; Gm728
Gene Symbols DNAI2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAI2. Recombinant protein control fragment (Product #RP-103079).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64446
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.