Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJA1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501567919
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7919 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7919 Supplier Bethyl Laboratories, Inc Supplier No. A304516AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan

It is a novel human homolog of E. coli′s DNAJ protein from heat shock protein 40 family. Hsp40 is involved in chaperonic protein folding HDJ-2 is cytoplasmic and nuclear membrane associated protein; upon heat shock it migrates and becomes associated with the Golgi complex, nucleolus and nuclear membrane HDJ-2 has a prenylated C-terminus and is not inducible upon heat shock.

Specifications

Antigen DNAJA1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene DNAJA1
Gene Accession No. P31689, P63037
Gene Alias HSJ-2, dnaJ homolog subfamily A member 1, dnaJ protein homolog 2, heat shock 40 kDa protein 4, heat shock protein J2, heat shock protein, DNAJ-like 2, human DnaJ protein 2, neural precursor cell expressed, developmentally down-regulated 7
Gene Symbols DNAJA1
Host Species Rabbit
Immunogen Between 250 and 300
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15502, 3301
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.