Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJA2 Antibody (OTI3A10), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271565AF350
Description
DNAJA2 Monoclonal specifically detects DNAJA2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
DNAJA2 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
DNAJA2 | |
Full length recombinant protein of human DNAJA2 (NP_005871) produced in HEK293T cells. | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3A10 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
cell cycle progression 3 protein, Cell cycle progression restoration gene 3 protein, CPR3PRO3015, Dj3, DJA2, DNAJ, DnaJ (Hsp40) homolog, subfamily A, member 2, dnaJ homolog subfamily A member 2, DNJ3, HIRA interacting protein 4, HIRA-interacting protein 4, HIRIP4RDJ2, Renal carcinoma antigen NY-REN-14 | |
Mouse | |
0.1 mL | |
10294 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction