Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJA2/CPR3 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13023R

Description
The protein encoded by this gene belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain. The product of this gene works as a cochaperone of Hsp70s in protein folding and mitochondrial protein import in vitro.Specifications
DNAJA2/CPR3 | |
Polyclonal | |
Unconjugated | |
DNAJA2 | |
1500017M13Rik; 2010206B19Rik; cell cycle progression 3 protein; Cell cycle progression restoration gene 3 protein; CPR3; Dj3; DJA2; DNA J protein; DNAJ; DnaJ (Hsp40) homolog, subfamily A, member 2; DnaJ heat shock protein family (Hsp40) member A2; DnaJ homolog subfamily A member 2; Dnaj3; Dnaja2; Dnj3; HIRA interacting protein 4; HIRA-interacting protein 4; HIRIP4; mDj3; PRO3015; RDJ2; Renal carcinoma antigen NY-REN-14 | |
Rabbit | |
Protein A | |
RUO | |
10294 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O60884 | |
DNAJA2 | |
KLH conjugated synthetic peptide derived from human DNAJA2/CPR3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction