Learn More
DNAJA2 Mouse anti-Canine, Human, Rat, Clone: OTI1G3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501709

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain. The product of this gene works as a cochaperone of Hsp70s in protein folding and mitochondrial protein import in vitro.Specifications
DNAJA2 | |
Monoclonal | |
Unconjugated | |
DNAJA2 | |
CPR3, DJ3, DJA2, DNAJ, DNJ3, HIRIP4, PRO3015, RDJ2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10294, 478143, 84026 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1G3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O35824, O60884 | |
Dnaja2 | |
Full length human recombit protein of human DNAJA2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.