Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530374
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530374 Supplier Invitrogen™ Supplier No. PA530374
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, Jurkat. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (88%), Xenopus laevis (93%), Chicken (96%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain. The product of this gene works as a cochaperone of Hsp70s in protein folding and mitochondrial protein import in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJA2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene DNAJA2
Gene Accession No. O60884
Gene Alias 1500017M13Rik; 2010206B19Rik; cell cycle progression 3 protein; cell cycle progression restoration gene 3 protein; CPR3; DJ3; DJA2; DNA J protein; DNAJ; DnaJ (Hsp40) homolog, subfamily A, member 2; DnaJ heat shock protein family (Hsp40) member A2; dnaJ homolog subfamily A member 2; Dnaj3; Dnaja2; DNJ3; EGK_12748; HIRA interacting protein 4; HIRA-interacting protein 4; HIRIP4; mDj3; PRO3015; Rdj2; Renal carcinoma antigen NY-REN-14
Gene Symbols DNAJA2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DNAJA2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.