Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565311 Supplier Invitrogen™ Supplier No. PA565311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGILIIQFLVI FPEKHWLSLE KLPQLEALLP PRQKVRITDD MDQVELKEFC PNEQNWRQHR EAYEEDEDGP QAGVQCQTA Highest antigen sequence identity to the following orthologs - mouse 86%, rat 91%.

DNAJA4 (DnaJ Heat Shock Protein Family (Hsp40) Member A4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include unfolded protein binding and heat shock protein binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJA4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJA4
Gene Accession No. Q8WW22
Gene Alias 1110021L12Rik; AV358213; Dj4; DnaJ (Hsp40) homolog, subfamily A, member 4; DnaJ heat shock protein family (Hsp40) member A4; dnaJ homolog subfamily A member 4; DNAJA4; heat shock protein, DNAJ-like 4; Hsj4; mmDjA4; MST104; MSTP104; PRO1472
Gene Symbols DNAJA4
Host Species Rabbit
Immunogen Recombinant Human DNAJA4. Recombinant protein control fragment (Product #a href=.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55466
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.