Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570301 Supplier Invitrogen™ Supplier No. PA570301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 86%; Zebrafish: 93%.

DNAJA4 (DnaJ Heat Shock Protein Family (Hsp40) Member A4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include unfolded protein binding and heat shock protein binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJA4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DNAJA4
Gene Accession No. 0
Gene Alias 1110021L12Rik; AV358213; Dj4; DnaJ (Hsp40) homolog, subfamily A, member 4; DnaJ heat shock protein family (Hsp40) member A4; dnaJ homolog subfamily A member 4; DNAJA4; heat shock protein, DNAJ-like 4; Hsj4; mmDjA4; MST104; MSTP104; PRO1472
Gene Symbols DNAJA4
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of rat DNAJA4.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300721
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.