Learn More
DNAJB1/Hsp40 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | DNAJB1/Hsp40 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-9914
![]() |
Bethyl Laboratories, Inc
A305245AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
Interacts with HSP70 and can stimulate its ATPase activity. Stimulates the association between HSC70 and HIP.Specifications
DNAJB1/Hsp40 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
DnaJ (Hsp40) homolog; subfmaily B; member 1; dnaJ homolog subfamily B member 1; dnaJ protein homolog 1; hDj-1; heat shock 40 kDa protein 1; heat shock 40kD protein 1; heat shock protein 40; human DnaJ protein 1; radial spoke 16 homolog B | |
DNAJB1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P25685 | |
3337 | |
Between 290 and 340 | |
Primary | |
4° C | |
DNAJB1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.