Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJB1 Mouse anti-Human, Clone: OTI1E10, lyophilized, TrueMAB™
SDP

Catalog No. 501661183
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-1183 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-1183 Supplier OriGene Supplier No. CF502262
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Interacts with HSP70 and can stimulate its ATPase activity. Stimulates the association between HSC70 and HIP.

Specifications

Antigen DNAJB1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1E10
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene DNAJB1
Gene Accession No. P25685
Gene Alias HSPF1, Hdj1, Hsp40, RSPH16B, Sis1
Gene Symbols DNAJB1
Host Species Mouse
Immunogen Full length human recombit protein of human DNAJB1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3337
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.